missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems FITC-Labeled Human BCMA / TNFRSF17 Protein, His Tag DMF Filed
4843.48 NOK - 33298.92 NOK
Specifications
Accession Number | NP_001183.2 |
---|---|
Molecular Weight (g/mol) | 7.8 kDa |
Source | FITC-Labeled Human BCMA, His Tag (Cat. No. BCA-HF2H1) is expressed from human HEK293 cells. It contains AA Met 1 - Ala 54 (Accession # Q02223-1). It is the FITC labeled form of Human BCMA, His Tag (Cat. No. BCA-H522y). |
Immunogen | Met 1 - Ala 54 |
Storage Requirements | -20°C, avoid light |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16447219
|
ACROBiosystems
BCA-HF2H1-25UG |
25 μg |
4843.48 NOK
25µg |
Estimated Shipment: 11-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16437219
|
ACROBiosystems
BCA-HF2H1-200UG |
200 μg |
33298.92 NOK
200µg |
Estimated Shipment: 11-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Tumor necrosis factor receptor superfamily member 17 (TNFRSF17) is also known as B-cell maturation protein (BCMA), CD antigen CD269, which is a member of the TNF-receptor superfamily. TNFRSF17 contains one TNFR-Cys repeat. TNFRSF17 is expressed in mature B-cells, but not in T-cells or monocytes. TNFRSF17 is receptor for TNFSF13B/BLyS/BAFF and TNFSF13/APRIL. TNFRSF17 promotes B-cell survival and plays a role in the regulation of humoral immunity. TNFRSF17 can activate NF-kappa-B and JNK.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 7.8 kDa. The protein migrates as 10 kDa and 14-16 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifications
NP_001183.2 | |
FITC-Labeled Human BCMA, His Tag (Cat. No. BCA-HF2H1) is expressed from human HEK293 cells. It contains AA Met 1 - Ala 54 (Accession # Q02223-1). It is the FITC labeled form of Human BCMA, His Tag (Cat. No. BCA-H522y). | |
-20°C, avoid light | |
BCMA | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Fluorescent labeled protein |
7.8 kDa | |
Met 1 - Ala 54 | |
TNFRSF17,CD269,BCM,BCMA | |
FITC | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |