Learn More
Fisherbrand™ Clear Standard Wide Mouth Bottles with White Polypropylene Pulp/Vinyl Lined Cap
Catalog Number p-3107646
Click to view available options
Capacity (Metric):
120 mL
240 mL
960 mL
Quantity:
12/Cs.
24/Cs.
General purpose bottle. Fisherbrand™ Clear Standard Wide Mouth Bottles with White Polypropylene Pulp/Vinyl Lined Cap are used for pharmaceutical, chemical, environmental, and food packaging and storing applications.
Clear / Flint Standard Wide Mouth Bottles feature a wide opening making them the ideal general purpose bottle. Applications include pharmaceutical, chemical, environmental, and food packaging and storing.
  • Features a wide mouth opening for easy transfer of contents
  • Applications include pharmaceutical, chemical, environmental, and food packaging and storing
  • Manufactured from clear flint glass
Capacity 4 oz., 125mL
Closure Liner Poly/Vinyl
Closure Material Polypropylene
Closure Size 48-400 mm
Closure Type Screw Cap
Diameter (Metric) 51 mm
Through the Closure Dispensing Yes
Product Type Standard Wide Mouth Bottle
Capacity (Metric) 120 mL
Capacity (English) 4 oz.
Color Clear
Material Glass
Quantity 24/Cs.
Closure Color White
Liner Material Pulp/Vinyl
Neck Finish 48-400
Closure Included Yes
Mouth Wide
Shape Round