Learn More
Fisherbrand™ 16-Tooth Combs for Horizontal Midigel Systems
16-Tooth Combs for Horizontal Midigel Systems
625.00 NOK - 650.00 NOK
Specifications
For Use With (Equipment) | Owl™ A1 Large Gel Electrophoresis System |
---|---|
Type | Standard Comb |
Teeth Count | 16 |
Width (English) Teeth | 0.24 in. |
Width (Metric) Teeth | 0.61 cm |
Product Code | Brand | Thickness (Metric) Teeth | Volume (Fill) Metric | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Thickness (Metric) Teeth | Volume (Fill) Metric | Price | Quantity & Availability | ||||
12433637
|
Fisherbrand™
|
0.1 cm | 30 μL |
625.00 NOK
Each |
Estimated Shipment: 16-07-2024
Log in to see stock. |
||||
12443637
|
Fisherbrand™
|
0.15 cm | 45 μL |
650.00 NOK
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
||||
Description
- Accessory for use with Horizontal Midigel Systems
Specifications
Owl™ A1 Large Gel Electrophoresis System | |
16 | |
0.61 cm |
Standard Comb | |
0.24 in. |