Learn More
ROBU VitraPOR™ Filter Disc, Surfaces Plane, Ground Cylindrical, 300 mm
The faces of the VitraPOR™ biplane filter discs from series 16 are precisely ground down to tight tolerances and plane surfaces.
14300.00 NOK - 17165.00 NOK
Specifications
Material | Borosilicate Glass (3.3) |
---|---|
Sterility | Non-sterile |
Thickness (Metric) | 18.0 mm ± 0.5 mm |
Diameter (Metric) Membrane | 300 mm |
Product Code | Brand | Porosity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Porosity | Price | Quantity & Availability | |||||
16863286
|
ROBU VitraPOR™
63000 |
0 |
14300.00 NOK
Each |
Estimated Shipment: 25-04-2024
Log in to see stock. |
|||||
16873286
|
ROBU VitraPOR™
63001 |
1 |
14300.00 NOK
Each |
Estimated Shipment: 25-04-2024
Log in to see stock. |
|||||
16883286
|
ROBU VitraPOR™
63002 |
2 |
14300.00 NOK
Each |
Estimated Shipment: 25-04-2024
Log in to see stock. |
|||||
16893286
|
ROBU VitraPOR™
63003 |
3 |
14300.00 NOK
Each |
Estimated Shipment: 25-04-2024
Log in to see stock. |
|||||
16803296
|
ROBU VitraPOR™
63004 |
4 |
17165.00 NOK
Each |
Estimated Shipment: 25-04-2024
Log in to see stock. |
|||||
Description
- Highly resistant to most chemicals and temperatures up to 500°C
- The biplane version may be required in specific installation circumstances
Specifications
Borosilicate Glass (3.3) | |
Non-sterile | |
18.0 mm ± 0.5 mm | |
300 mm |