Learn More
ROBU™ by STEREON VitraPOR™ Porous Glass Filter Candle Blank, Conical Shape, ø 34 mm
Product Code. 10002081
Change view
Click to view available options
Pore Size:
10 to 16 μm
100 to 160 μm
16 to 40 μm
160 to 250 μm
40 to 100 μm
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size Unit Size
10002081 100 to 160 μm Each
10092071 160 to 250 μm Each
10194401 40 to 100 μm Each
10574351 16 to 40 μm Each
10584351 10 to 16 μm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
When working with glassware always wear protective glasses and protective gloves to avoid injuries. Use a safety screen, -hood or similar protection when working with glassware subject to pressure or vacuum. Be prepared that reagents may leak from a broken vessel. Please heed these guidelines in combination with the respective state-specific regulations for the use of glasware in the laboratory.
Product Code. 10002081 Supplier ROBU™ by STEREON Supplier No. 18211
Estimated Shipment: 15-12-2026
When working with glassware always wear protective glasses and protective gloves to avoid injuries. Use a safety screen, -hood or similar protection when working with glassware subject to pressure or vacuum. Be prepared that reagents may leak from a broken vessel. Please heed these guidelines in combination with the respective state-specific regulations for the use of glasware in the laboratory.
Designed for OEM filtration, gas diffusion and custom laboratory or process assemblies using borosilicate 3.3 glass.
  • Supplied without an attached glass tube
  • Available in conical and cylindrical geometries
  • Multiple sizes and porosity classes cover coarse gas distribution through fine filtration
  • Custom housings and probes
  • Chemically resistant all-glass filter medium with no separate support mesh
  • Suitable for custom component development and replacement of legacy porous glass elements
Specifications
Autoclavable Autoclavable
Disposable Single-use
Filter Type Air/Gas/Solvent
Format Cylindrical Filter Candle
Material Borosilicate Glass (3.3)
Material (Membrane) Glass Fiber
Pore Size 100 to 160 μm
Sterility Non-sterile
Max. Pressure 1 bar
Porosity 1
Type Filter Candle
For Use With (Equipment) VitraPOR
Show More Show Less
Nominal dimensions and technical data may vary within manufacturing tolerances. Validate chemical, thermal and mechanical suitability for the intended application.