missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Stedim Biotech™ Filter Boards, Discs, Grade 157
Filter boards for the filtration and absorption of liquids.
Brand: Sartorius Stedim Biotech™ FT-3-437-022
Additional Details : Weight : 0.26600kg
Description
- Constructed of cotton linters and refined pulp
- Suitable for analytical sample preparation
- Non-sterile, bulk packed
Specifications
700 g/m2 | |
Cotton Linters, Refined Pulp | |
1000/Pk. | |
22 mm |
Disc | |
157 | |
1.8 mm |