Learn More
Gibco™ FGF-Basic Full Length Recombinant Human Protein
Recombinant Human FGF-basic (full length)
10420.00 NOK - 21560.00 NOK
Specifications
For Use With (Application) | Cell Culture |
---|---|
Molecular Weight (g/mol) | 17.2 kDa |
Purification Method | Sequential Chromatography |
Gene Alias | FGF |
Protein Length | 155 aa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11390832
|
Gibco™
CTP0261 |
100 μg |
10420.00 NOK
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11300842
|
Gibco™
CTP0263 |
1 mg |
21560.00 NOK
Each |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Cell Therapy Systems CTS™ Recombinant Human FGF-basic (full length) is a bioactive protein intended for use in cell culture applications, including pre-clinical and clinical cell therapy studies. FGF-basic (FGFb), is involved in a number of biological processes including embryonic development and differentiation, and neuron differentiation, survival, and regeneration. FGFb is also involved in the proliferation of cells of mesodermal origin and many cells of neuroectodermal, ectodermal, and endodermal origin. The lyophilized, E. coli-expressed FGF-basic (full length) CTS™ Recombinant Human is verified for activity through dose-dependent proliferation of BALB/3T3 cells.
- Qualified—helps reduce your burden in qualifying reagents for regulatory submission in cell therapy applications
- Highly Pure—minimizes interference from other proteins or contaminants
- Active—get more results with less protein
Simplified Transition into Clinical Research
GIBCO™ Cell Therapy Systems (CTS™) products are of high quality and supplied with harmonized documentation such as Certificates of Analysis, Certificates of Origin, and access to authorization letters for our Drug Master Files, as appropriate. The GIBCO™ CTS™ product line enables you to reduce your burden in qualifying reagents during your transition from research applications to clinical applications.
High Purity
To help verify that GIBCO™ CTS™ Recombinant growth factors are of the highest quality, each protein is analyzed for purity, along with structural homogeneity, to help ensure a biologically active protein. The purity of each GIBCO™ CTS™ recombinant growth factor is verified by SDS-PAGE to be over 95% pure and to have endotoxin levels below 0.1 ng/μg.
High Biological Activity
All GIBCO™ CTS™ Recombinant growth factors are verified for activity. Each lot has its own ED50 determined using an activity assay based on a specific function of the growth factor. The activity of our FGF-basic (full length) CTS™ Recombinant Human growth factor is determined by the dose-dependent proliferation of BALB/3T3 cells to have an ED50 range of 0.1–1.0 ng/mL (specific activity: 1.0 × 107–1.0 × 106 units/mg). Optimal concentration for individual applications should be determined by a dose response assay.
Specifications
Cell Culture | |
Sequential Chromatography | |
155 aa | |
Recombinant | |
Carrier-Free | |
Proliferation of BALB⁄3T3 cells | |
Wet Ice | |
Growth Factors & Receptors | |
Lyophilized | |
Clinical Research, Oncology, Angiogenesis, Stem Cell Research, Microbiology, Differentiation, Neurobiology | |
95 % |
17.2 kDa | |
FGF | |
Human | |
2°C to 8°C | |
0.1 ng/ml | |
<0.1 ng/μg | |
E. coli | |
Full Length, Recombinant, Ligand | |
FGFs (Fibroblast Growth Factors) | |
Gibco™ |
For Research Use Only. Not for use in diagnostic procedures.