Learn More
Ferric Ammonium Sulphate R2 solution EU Pharmacopoeia, Fisher Chemical™
Catalog Number 12950414
Click to view available options
Quantity:
1000 mL
Packaging:
Ampoule
Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.
CAS 74738-17-3
Quantity 1000 mL
Molecular Formula C11H6Cl2N2
Synonym fenpiclonil, beret, 3-2,3-dichlorophenyl-4-cyanopyrrole, fenpiclonil iso, 4-2,3-dichlorophenyl-1h-pyrrole-3-carbonitrile, unii-21ocb8kdq9, 4-cyano-3-2,3-dichlorophenyl pyrrole, 21ocb8kdq9, 1h-pyrrole-3-carbonitrile, 4-2,3-dichlorophenyl, 4-2,3-dichlorophenyl pyrrole-3-carbonitrile
InChI Key FKLFBQCQQYDUAM-UHFFFAOYSA-N
SMILES C1=CC(=C(C(=C1)Cl)Cl)C2=CNC=C2C#N
IUPAC Name 4-(2,3-dichlorophenyl)-1H-pyrrole-3-carbonitrile
Molecular Weight (g/mol) 237.083
PubChem CID 91724
ChEBI CHEBI:34759
Concentration or Composition (by Analyte or Components) 10μg/mL
Packaging Ampoule
Grade EU Ph Ref: 1037702
Show More Show Less
compliance-icons
Product Identifier
  • Ferric ammonium sulphate solution R2. European Pharmacopoeia Ref: 1037702 (RP 077)
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.