missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ FBXO39 Recombinant Protein

4115.00 NOK - 6245.00 NOK

Specifications

Accession Number AAH34782
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
Gene ID (Entrez) 162517
Molecular Weight (g/mol) 74.14
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16128943
View Documents Promotion Details
Abnova™
H00162517-P01.10UG
10 ug
4115.00 NOK
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 11-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16138943
View Documents Promotion Details
Abnova™
H00162517-P01.25UG
25 ug
6245.00 NOK
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 11-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH34782
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
74.14
8
Glutathione Sepharose 4 Fast Flow
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FBXO39
Human
Recombinant
Solution
Antibody Production, Protein Array, ELISA, Western Blot
162517
FBXO39 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
MDEESELIQPQDQSCWAFLPDLCLCRVFWWLGDRDRSRAALVCRKWNQMMYSAELWRYRTITFSGRPSRVHASEVESAVWYVKKFGRYLEHLEVKFMNPYNAVLTKKFQVTMRGLLSCLSKSNNRLKSLSIQYLELDRLVWRNSIRSSFISSLSFFLKKMGKRLDYLNLKGARLTVEQGCQILDSLSYMRNENVISELNIEDYFSHHLAVYNSPQFKKTMSTFHNLVSLNLNYNCISDELLENLCENASTLRTINIKCHVHDPHGQVIWGMSWAKLARQATNLKVNFFFERIMKYERLARILLQEIPIRSISLRSCYFSDPDCSMRPTLIDLLPTFRHTLQKLTCEFNNNHESLDEELHLLIISCRKLFYFKIWAFLDVSFVERILKSQKERQCALRVFKARIYTNRYETNEEDKTLQEIYRKYRKLIESELSYFVIVYSVM
Fbx39/MGC35179
FBXO39
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Product Certifications