Learn More
Fisherbrand™ FB65464 Rotary Vane Vacuum Pump
Two-stage rotary vane vacuum pump. Fisherbrand™ FB65464 Rotary Vane Vacuum Pump designed for laboratory applications requiring a deep vacuum.
Brand:  Fisherbrand™
Additional Details : Weight : 35.00000kg
Product Code. 12951141
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
These two stage rotary vane pumps provide a high water vapour tolerance. The low weight, small footprint and high pumping speed, together with increased chemical resistance, are ideal features for both research and routine applications.
  • Compact design, low weight and easy to use
  • Quiet running and excellent ultimate vacuum
  • Active corrosion protection
  • Easy maintenance
  • Supplied with clamps and seals for the inlet and outlet
  • Each pumps comes with anti-suck back valve, gas ballast valve, oil charging, centring ring, clamping ring and motor overload protection For a wide range of laboratory applications and continuous running.
Specifications
Specifications
2 x 10-3 mbar
115/230 V 50/60 Hz motor with Schuko UK and US power leads
170 mm
35 kg
260 mm
Rotary Vane Vacuum Pump
100/230 V
183/220 L/min.
54 x 17 x 26 cm (21 x 6.69 x 10.2 in.)
FB65464
50/60 Hz
540 mm
183 L/min.