Learn More
DWK Life Sciences Wheaton™ CELLine™ Bioreactor Flask
Membrane cell culture flasks ideal for antibody and protein production
2965.00 NOK - 16120.00 NOK
Specifications
Includes | User manual with feeding and harvesting schedules |
---|---|
Sterility | Sterile |
Product Code | Brand | Capacity (Metric) Cell | Capacity (Metric) Media | For Use With (Application) | No. per Case | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) Cell | Capacity (Metric) Media | For Use With (Application) | No. per Case | Price | Quantity & Availability | ||
12871793
|
DWK Life Sciences
WCL1000-3 |
15 mL | 1000 mL | Membrane cell culture flasks ideal for antibody and Protein production, for suspension cultures | 3/Cs. |
15025.00 NOK
Pack of 3 |
Estimated Shipment: 09-08-2024
Log in to see stock. |
||
12801803
|
DWK Life Sciences
WCL1000-1 |
15 mL | 1000 mL | Membrane cell culture flasks ideal for antibody and Protein production, for suspension cultures | 1/Cs. |
3635.00 NOK
Each |
Estimated Shipment: 22-05-2024
Log in to see stock. |
||
12891793
|
DWK Life Sciences
WCL1000AD-1 |
15 mL | 1000 mL | Membrane cell culture flasks ideal for antibody and Protein production, for adherent cultures | 1/Cs. |
3820.00 NOK
Each |
Estimated Shipment: 07-05-2024
Log in to see stock. |
||
12881793
|
DWK Life Sciences
WCL0350-5 |
5 mL | 350 mL | Membrane cell culture flasks ideal for antibody and Protein production, for suspension cultures | 5/Cs. |
16120.00 NOK
Pack of 5 |
Estimated Shipment: 18-09-2024
Log in to see stock. |
||
12811803
|
DWK Life Sciences
WCL1000AD-3 |
15 mL | 1000 mL | Membrane cell culture flasks ideal for antibody and Protein production, for adherent cultures | 3/Cs. |
11450.00 NOK
Pack of 3 |
Estimated Shipment: 07-05-2024
Log in to see stock. |
||
12821803
|
DWK Life Sciences
WCL0350-1 |
5 mL | 350 mL | Membrane cell culture flasks ideal for antibody and Protein production, for suspension cultures | 1/Cs. |
2965.00 NOK
Each |
Estimated Shipment: 07-05-2024
Log in to see stock. |
||
Description
High-density cell culture flasks with dual membrane system allow for less culture maintenance and a high concentration harvest.
- Bulk media chamber allows for longer run times without media refreshing
- Cell compartment concentrates cells, antibodies and proteins for simplified purification
- For suspension cell culture and adherent cell culture: Hybridoma, CHO, NS0, SF, BHK, HEK, etc.
02-912-745, 02-912-742
- For adherent cell cultivation
- 1000 mL media compartment
- 15 mL cell compartment
- Available as package of one (02-912-745) or package three (02-912-742)
- For suspension cell cultivation
- 350 mL media compartment
- 5 mL cell compartment
- Available as package of one (02-912-746) or package five (02-912-746)
- For suspension cell cultivation
- 1000 mL media compartment
- 15 mL cell compartment
- Available as package of one (02-912-744) or package three (02-912-741)
Specifications
User manual with feeding and harvesting schedules | |
Sterile |