missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Factor V Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
2755.00 NOK - 4535.00 NOK
Specifications
| Antigen | Factor V |
|---|---|
| Dilution | Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18463321
|
Novus Biologicals
NBP1-88114-25ul |
25 μL |
2755.00 NOK
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18252828
|
Novus Biologicals
NBP1-88114 |
0.1 mL |
4535.00 NOK
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Factor V Polyclonal specifically detects Factor V in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| Factor V | |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| Activated protein C cofactor, coagulation factor V, coagulation factor V (proaccelerin, labile factor), coagulation factor V jinjiang A2 domain, factor V Leiden, FVL, PCCF, Proaccelerin, labile factor | |
| F5 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| P12259 | |
| 2153 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:MPSPSSPTLNDTFLSKEFNPLVIVGLSKDGTDYIEIIPKEEVQSSEDDYAEIDYVPYDDPYKTDVRTNINSSRDPDNIAAWYLRSNNGNRRNYYIAAEEISWDYSEFVQRETDIEDSDDIPEDTT | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title