Learn More
F(ab')2-Goat anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor™ Plus 488, Invitrogen™
F(ab')2-Goat Polyclonal Secondary Antibody
2120.00 NOK - 3345.00 NOK
Specifications
Antigen | Rabbit IgG (H+L) Cross-Adsorbed |
---|---|
Concentration | 2 mg/mL |
Applications | Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Frozen) |
Classification | Polyclonal |
Conjugate | Alexa Fluor Plus 488 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17226553
|
Invitrogen
A48282TR |
100 μg |
2120.00 NOK
100µg |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17218370
|
ABI Applied Biosystems
A48282 |
500 μg |
3345.00 NOK
500µg |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
F(ab')2 antibody fragments offer several benefits over whole IgGs, due to their smaller size (∽110 kD vs. ∽ 150 kD) for better penetration into tissue sections, resulting in improved staining in applications such as IHC. The lack of Fc region results in reduced nonspecific background staining when staining cells with Fc receptors (such as immune cells) and other Fc interaction-related interference. F(ab')2 antibody fragments were obtained by a combination of pepsin digestion and immunoaffinity chromatography to remove residual Fc fragments and undigested whole IgG molecules. To minimize cross-reactivity, the goat anti-rabbit IgG F(ab')2 fragments have been cross-adsorbed against human, mouse and rat serum proteins to increase specificity of the antibody resulting in higher sensitivity and less background staining. This antibody binds to heavy chains on rabbit IgG and light chains on all rabbit immunoglobulins, does not bind to human, mouse or rat serum proteins/IgG or rabbit non-immunoglobulin serum proteins.
Rabbit IgG (H+L) Cross-Adsorbed Polyclonal antibody specifically detects Rabbit IgG (H+L) Cross-Adsorbed in Rabbit samples. It is validated for Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Frozen).Specifications
Rabbit IgG (H+L) Cross-Adsorbed | |
Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
Alexa Fluor Plus 488 | |
Goat | |
Rabbit | |
Gamma Immunoglobins Heavy and Light chains | |
Secondary | |
4° C, store in dark |
2 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
proprietary buffer with 0.016% Bromonitrodioxane, 0.016% Methylisothiazolone; pH 6.5 | |
IgG | |
Affinity chromatography |