Learn More
F-15599, MedChemExpress
Product Code. 16460408
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
20 mg
Unit Size:
10mg
1mg
20mg
5mg
Product Code. 16460408
Brand: Medchem Express HY19863
MedChemExpress F-15599 is a highly selective G-protein biased 5-HT1A receptor agonist, with Ki of 3.4 nM.
F-15599 is a highly selective G-protein biased 5-HT1A receptor agonist, with Ki of 3.4 nM.
Specifications
Chemical Name or Material F-15599
CAS 635323-95-4
Color Off-White
Purity Grade Notes Research
Molecular Formula C19H21ClF2N4O
Quantity 5 mg
Synonym NLX-101
Solubility Information DMSO : 250 mg/mL (633.15 mM; Need ultrasonic)
SMILES O=C(C1=CC=C(F)C(Cl)=C1)N2CCC(CNCC3=NC=C(C)C=N3)(F)CC2
Molecular Weight (g/mol) 394.85
Formula Weight 394.85
Percent Purity 99.37%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.