Learn More
EVOS™ EVOS™ 4X Objective, fluorite, LWD, phase-contrast
Components manufactured to ensure publication-quality images
Brand: EVOS™ AMEP4680
Additional Details : Weight : 0.10000kg
Description
RMS-threaded objectives with 45mm parfocal distance
Specifications
EVOS LWD Fluorite Phase Contrast Objective | |
EVOS™ XL Core Imaging System, EVOS™ FL Color Imaging System, EVOS™ XL Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System | |
4X | |
1 Ea. |
EVOS | |
Fluorite | |
0.13 |
For Research Use Only. Not for use in diagnostic procedures.