Learn More
Ethylhydrocupreine hydrochloride, 97%, Thermo Scientific Chemicals
Catalog Number p-7025900
Click to view available options
Quantity:
100 mg
500 mg
3413-58-9
C21H29ClN2O2
376.925
MFCD00135594
QNRATNLHPGXHMA-XZHTYLCXSA-N
ethylhydrocupreine hydrochloride, optoquin hydrochloride, ethylhydrocupreine hcl, optochin hydrochloride, unii-s3n0jb4fc7, neumolisina, numoquin hydrochloride, s3n0jb4fc7, optoquinhydrochloride, optochin
16219340
(R)-(6-ethoxyquinolin-4-yl)-[(2S,4S,5R)-5-ethyl-1-azabicyclo[2.2.2]octan-2-yl]methanol;hydrochloride
CCC1CN2CCC1CC2C(C3=C4C=C(C=CC4=NC=C3)OCC)O.Cl
3413-58-9
C21H29ClN2O2
376.925
MFCD00135594
QNRATNLHPGXHMA-XZHTYLCXSA-N
ethylhydrocupreine hydrochloride, optoquin hydrochloride, ethylhydrocupreine hcl, optochin hydrochloride, unii-s3n0jb4fc7, neumolisina, numoquin hydrochloride, s3n0jb4fc7, optoquinhydrochloride, optochin
16219340
(R)-(6-ethoxyquinolin-4-yl)-[(2S,4S,5R)-5-ethyl-1-azabicyclo[2.2.2]octan-2-yl]methanol;hydrochloride
CCC1CN2CCC1CC2C(C3=C4C=C(C=CC4=NC=C3)OCC)O.Cl
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point ∼240°C (decomposition)
Quantity 100 mg
Sensitivity Light sensitive
Formula Weight 374.91
Percent Purity 97%
Chemical Name or Material Ethylhydrocupreine hydrochloride