Learn More
Ethyl nipecotate, 97%, Thermo Scientific Chemicals
Catalog Number p-3736495
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
5006-62-2
C8H15NO2
157.21
MFCD00005991
XIWBSOUNZWSFKU-UHFFFAOYSA-N
ethyl nipecotate, ethyl 3-piperidinecarboxylate, nipecotic acid ethyl ester, 3-piperidinecarboxylic acid ethyl ester, ethylpiperidine-3-carboxylate, 3-ethoxycarbonyl piperidine, 3-piperidinecarboxylic acid, ethyl ester, piperidine-3-carboxylic acid ethyl ester, ethylnipecotate, a-ethyl nipecotate
98969
ethyl piperidine-3-carboxylate
CCOC(=O)C1CCCNC1
5006-62-2
C8H15NO2
157.21
MFCD00005991
XIWBSOUNZWSFKU-UHFFFAOYSA-N
ethyl nipecotate, ethyl 3-piperidinecarboxylate, nipecotic acid ethyl ester, 3-piperidinecarboxylic acid ethyl ester, ethylpiperidine-3-carboxylate, 3-ethoxycarbonyl piperidine, 3-piperidinecarboxylic acid, ethyl ester, piperidine-3-carboxylic acid ethyl ester, ethylnipecotate, a-ethyl nipecotate
98969
ethyl piperidine-3-carboxylate
CCOC(=O)C1CCCNC1
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Density 1.0100g/mL
Boiling Point 102°C to 104°C (7.0 mmHg)
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4580 to 1.461
Quantity 25 g
Specific Gravity 1.01
Solubility Information Solubility in water: miscible. Other solubilities: soluble in most organic solvents
Formula Weight 157.21
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Ethyl nipecotate
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl nipecotate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.