Learn More
1,2-Bis(diphenylphosphino)ethane, 98+%, Thermo Scientific Chemicals
Catalog Number p-129136
Click to view available options
Quantity:
1 g
10 g
50 g
250 g
Packaging:
Glass bottle
1663-45-2
C26H24P2
398.43
MFCD00003047
QFMZQPDHXULLKC-UHFFFAOYSA-N
1,2-bis diphenylphosphino ethane, diphos, ethylenebis diphenylphosphine, bis diphenylphosphine ethane, bis 1,2-diphenylphosphino ethane, phosphine, 1,2-ethanediylbis diphenyl, phosphine, ethylenebis diphenyl, 1,2-bis diphenylphosphine ethane, 1,2-bis diphenylphosphino-ethane, unii-kl33qe52i4
74267
CHEBI:30669
2-diphenylphosphanylethyl(diphenyl)phosphane
C(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
1663-45-2
C26H24P2
398.43
MFCD00003047
QFMZQPDHXULLKC-UHFFFAOYSA-N
1,2-bis diphenylphosphino ethane, diphos, ethylenebis diphenylphosphine, bis diphenylphosphine ethane, bis 1,2-diphenylphosphino ethane, phosphine, 1,2-ethanediylbis diphenyl, phosphine, ethylenebis diphenyl, 1,2-bis diphenylphosphine ethane, 1,2-bis diphenylphosphino-ethane, unii-kl33qe52i4
74267
CHEBI:30669
2-diphenylphosphanylethyl(diphenyl)phosphane
C(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 138.0°C to 142.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98% min. (HPLC)
Packaging Glass bottle
Linear Formula (C6H5)2PCH2CH2P(C6H5)2
Quantity 1 g
Merck Index 15,1254
Formula Weight 398.41
Percent Purity 98+%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 1, 2-Bis(diphenylphosphino)ethane, 98+%
Show More Show Less
compliance-icons
Product Identifier
  • 1,2-Bis(diphenylphosphino)ethane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.