missing translation for 'onlineSavingsMsg'
Learn More

ethyle2-(phenylthio)acetate, 97%, Thermo Scientific™

Catalog Number p-2703995
Click to view available options
Quantity:
1 g
10 g
50 g
This item is not returnable. View return policy
7605-25-6
C10H12O2S
196.264
MFCD00145107
SEDRTXNDGKRHBL-UHFFFAOYSA-N
ethyl phenylthio acetate, ethyl 2-phenylthio acetate, ethyl 2-phenylsulfanyl acetate, acetic acid, phenylthio-, ethyl ester, ethyl 2-phenylthioacetate, phenylthio acetic acid ethyl ester, phenylthio acetic acid, ethyl ester, ethyl phenylsulfanyl acetate, pubchem10853
97312
ethyl 2-phenylsulfanylacetate
CCOC(=O)CSC1=CC=CC=C1
This item is not returnable. View return policy
7605-25-6
C10H12O2S
196.264
MFCD00145107
SEDRTXNDGKRHBL-UHFFFAOYSA-N
ethyl phenylthio acetate, ethyl 2-phenylthio acetate, ethyl 2-phenylsulfanyl acetate, acetic acid, phenylthio-, ethyl ester, ethyl 2-phenylthioacetate, phenylthio acetic acid ethyl ester, phenylthio acetic acid, ethyl ester, ethyl phenylsulfanyl acetate, pubchem10853
97312
ethyl 2-phenylsulfanylacetate
CCOC(=O)CSC1=CC=CC=C1
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Linear Formula C6H5SCH2C(O)OCH2CH3
Quantity 1 g
Formula Weight 196.27
Percent Purity 97%
Chemical Name or Material Ethyl 2-(phenylthio)acetate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'