missing translation for 'onlineSavingsMsg'
Learn More

Ethyl vinyl ketone, 98%, stabilized, Thermo Scientific™

Catalog Number p-3736501
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1629-58-9, 128-37-0
C5H8O
84.12
MFCD00009316
JLIDVCMBCGBIEY-UHFFFAOYSA-N
1-penten-3-one, ethyl vinyl ketone, ethylvinylketone, 1-pentene-3-one, ethylvinyl ketone, pentenone, ketone, ethyl vinyl, vinyl ethyl ketone, unii-r0053y1az7, fema no. 3382
15394
pent-1-en-3-one
CCC(=O)C=C
This item is not returnable. View return policy
1629-58-9, 128-37-0
C5H8O
84.12
MFCD00009316
JLIDVCMBCGBIEY-UHFFFAOYSA-N
1-penten-3-one, ethyl vinyl ketone, ethylvinylketone, 1-pentene-3-one, ethylvinyl ketone, pentenone, ketone, ethyl vinyl, vinyl ethyl ketone, unii-r0053y1az7, fema no. 3382
15394
pent-1-en-3-one
CCC(=O)C=C

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Amber to Colorless
Density 0.8490g/mL
Boiling Point 103°C to 104°C
Flash Point -6°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C2H5COCH=CH2
Refractive Index 1.4180 to 1.422
Quantity 5 g
Beilstein 01, 731
Fieser 01,388
Specific Gravity 0.849
Stabilizer 0.1% (BHT)
Formula Weight 84.12
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl vinyl ketone, Stabilized
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl vinyl ketone, stabilized
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'