missing translation for 'onlineSavingsMsg'
Learn More

Ethyl methylsulfonylacetate, 97%, Thermo Scientific™

Catalog Number p-7044186
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
4455-15-6
C5H10O4S
166.191
MFCD00041258
OCCWQCYBCZADCE-UHFFFAOYSA-N
ethyl methanesulfonylacetate, ethyl 2-methylsulfonyl acetate, ethyl methylsulfonylacetate, ethyl methylsulphonyl acetate, ethyl methanesulphonylacetate, ethyl methylsulfonyl acetate, ethyl 2-methanesulfonylacetate, acetic acid, methylsulfonyl-, ethyl ester, ethylmethanesulfonylacetate
78201
ethyl 2-methylsulfonylacetate
CCOC(=O)CS(=O)(=O)C
This item is not returnable. View return policy
4455-15-6
C5H10O4S
166.191
MFCD00041258
OCCWQCYBCZADCE-UHFFFAOYSA-N
ethyl methanesulfonylacetate, ethyl 2-methylsulfonyl acetate, ethyl methylsulfonylacetate, ethyl methylsulphonyl acetate, ethyl methanesulphonylacetate, ethyl methylsulfonyl acetate, ethyl 2-methanesulfonylacetate, acetic acid, methylsulfonyl-, ethyl ester, ethylmethanesulfonylacetate
78201
ethyl 2-methylsulfonylacetate
CCOC(=O)CS(=O)(=O)C

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Density 1.26
Boiling Point 111°C to 113°C (0.2 mmHg)
Flash Point >100°C (212°F)
Refractive Index 1.457
Quantity 5 g
Beilstein 1771631
Formula Weight 166.2
Percent Purity 97%
Chemical Name or Material Ethyl methylsulfonylacetate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'