Learn More
Ethyl methyl sulfide, 97%, Thermo Scientific Chemicals
Catalog Number p-7043814
546.02 NOK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
624-89-5
C3H8S
76.16
MFCD00009268
WXEHBUMAEPOYKP-UHFFFAOYSA-N
ethyl methyl sulfide, methylthioethane, ethane, methylthio, methyl ethyl sulfide, ethyl methyl sulphide, methyl ethyl sulphide, sulfide, ethyl methyl, unii-mr1gzs62tm, methylthio ethane, methylsulfanyl ethane
12230
methylsulfanylethane
CCSC
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
624-89-5
C3H8S
76.16
MFCD00009268
WXEHBUMAEPOYKP-UHFFFAOYSA-N
ethyl methyl sulfide, methylthioethane, ethane, methylthio, methyl ethyl sulfide, ethyl methyl sulphide, methyl ethyl sulphide, sulfide, ethyl methyl, unii-mr1gzs62tm, methylthio ethane, methylsulfanyl ethane
12230
methylsulfanylethane
CCSC
Ethyl Methyl Sulfide is used as a corrosion inhibitor for zinc metal which finds wide applications like electrode material in batteries, sacrificial anodes and metallic coatings. It is used in mixtures designed to odorize natural gas and is made to by-pass injection system.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethyl Methyl Sulfide is used as a corrosion inhibitor for zinc metal which finds wide applications like electrode material in batteries, sacrificial anodes and metallic coatings. It is used in mixtures designed to odorize natural gas and is made to by-pass injection system.
Solubility
Miscible with alcohols and oils. Immiscible with water.
Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point -106°C
Density 0.841
Boiling Point 66°C to 67°C
Flash Point −15°C (5°F)
Odor Stench
Linear Formula CH3CH2SCH3
Refractive Index 1.4392
Quantity 25 g
UN Number UN1993
Beilstein 1696871
Solubility Information Miscible with alcohols and oils. Immiscible with water.
Formula Weight 76.16
Percent Purity 97%
Chemical Name or Material Ethyl methyl sulfide