missing translation for 'onlineSavingsMsg'
Learn More

Ethyl isocyanoacetate, 98%, Thermo Scientific Chemicals

Catalog Number p-7046629
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
2999-46-4
C5H7NO2
113.116
MFCD00000007
FPULFENIJDPZBX-UHFFFAOYSA-N
ethyl isocyanoacetate, isocyanoacetic acid ethyl ester, ethylisocyanoacetate, acetic acid, 2-isocyano-, ethyl ester, acetic acid, isocyano-, ethyl ester, ethyl iso-cyanoacetate, ethyl isocyano-acetate, pubchem2345, ethyl 2-isocyanoacetat
533707
ethyl 2-isocyanoacetate
CCOC(=O)C[N+]#[C-]
This item is not returnable. View return policy
2999-46-4
C5H7NO2
113.116
MFCD00000007
FPULFENIJDPZBX-UHFFFAOYSA-N
ethyl isocyanoacetate, isocyanoacetic acid ethyl ester, ethylisocyanoacetate, acetic acid, 2-isocyano-, ethyl ester, acetic acid, isocyano-, ethyl ester, ethyl iso-cyanoacetate, ethyl isocyano-acetate, pubchem2345, ethyl 2-isocyanoacetat
533707
ethyl 2-isocyanoacetate
CCOC(=O)C[N+]#[C-]

Ethyl isocyanoacetate is used in the syntheses of 7-aza-tetrahydroindoles, oxazolines, benzodiazepines, imidazoles and oxazoles. For example, it is involved in the synthesis of 1H-indole-2-carboxylate by condensing with 2-bromo benzenealdehyde using CuI as a catalyst. It is a precursor used in the preparation of ethyl thiazole-4-carboxylate upon reaction with O-ethyl thioformate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl isocyanoacetate is used in the syntheses of 7-aza-tetrahydroindoles, oxazolines, benzodiazepines, imidazoles and oxazoles. For example, it is involved in the synthesis of 1H-indole-2-carboxylate by condensing with 2-bromo benzenealdehyde using CuI as a catalyst. It is a precursor used in the preparation of ethyl thiazole-4-carboxylate upon reaction with O-ethyl thioformate.

Solubility
Miscible with organic solvents. Slightly miscible with water.

Notes
Store in cool place. Moisture and light sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY
Density 1.035
Boiling Point 194°C to 196°C
Flash Point 84°C (183°F)
Refractive Index 1.422
Quantity 1 g
UN Number UN2810
Beilstein 3588613
Sensitivity Light and moisture sensitive
Solubility Information Miscible with organic solvents. Slightly miscible with water.
Formula Weight 113.12
Percent Purity 98%
Chemical Name or Material Ethyl isocyanoacetate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'