missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl imidazole-4-carboxylate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
A ring-substituted-1H-imidazole-4-carboxylic acid derivative, that is shown to act as a new class of anti-tuberculosis agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
A ring-substituted-1H-imidazole-4-carboxylic acid derivative, that is shown to act as a new class of anti-tuberculosis agents.
Solubility
Soluble in dimethyl sulfoxide and methanol. Slightly soluble in water.
Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
A ring-substituted-1H-imidazole-4-carboxylic acid derivative, that is shown to act as a new class of anti-tuberculosis agents.
Solubility
Soluble in dimethyl sulfoxide and methanol. Slightly soluble in water.
Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
Specifications
Melting Point | 156°C to 160°C |
Quantity | 1 g |
Beilstein | 606937 |
Solubility Information | Soluble in dimethyl sulfoxide and methanol. Slightly soluble in water. |
Formula Weight | 140.14 |
Percent Purity | 98% |
Physical Form | Solid |
Chemical Name or Material | Ethyl imidazole-4-carboxylate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'