Learn More
Ethyl imidazole-4-carboxylate, 98%, Thermo Scientific Chemicals
Catalog Number p-7055496
Click to view available options
Quantity:
1 g
5 g
25 g
23785-21-9
C6H8N2O2
140.14
MFCD02646639,MFCD07363812
KLWYPRNPRNPORS-UHFFFAOYSA-N
ethyl 1h-imidazole-4-carboxylate, ethyl imidazole-4-carboxylate, ethyl 4-imidazolecarboxylate, ethyl 4 5-imidazolecarboxylate, 4-carbethoxyimidazole, 1h-imidazole-4-carboxylic acid ethyl ester, nic7lr0zon, unii-nic7lr0zon, imidazole-4-carboxylic acid ethyl ester, 1h-imidazole-4-carboxylic acid, ethyl ester
99170
ethyl 1H-imidazole-5-carboxylate
CCOC(=O)C1=CN=CN1
23785-21-9
C6H8N2O2
140.14
MFCD02646639,MFCD07363812
KLWYPRNPRNPORS-UHFFFAOYSA-N
ethyl 1h-imidazole-4-carboxylate, ethyl imidazole-4-carboxylate, ethyl 4-imidazolecarboxylate, ethyl 4 5-imidazolecarboxylate, 4-carbethoxyimidazole, 1h-imidazole-4-carboxylic acid ethyl ester, nic7lr0zon, unii-nic7lr0zon, imidazole-4-carboxylic acid ethyl ester, 1h-imidazole-4-carboxylic acid, ethyl ester
99170
ethyl 1H-imidazole-5-carboxylate
CCOC(=O)C1=CN=CN1
A ring-substituted-1H-imidazole-4-carboxylic acid derivative, that is shown to act as a new class of anti-tuberculosis agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
A ring-substituted-1H-imidazole-4-carboxylic acid derivative, that is shown to act as a new class of anti-tuberculosis agents.
Solubility
Soluble in dimethyl sulfoxide and methanol. Slightly soluble in water.
Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Melting Point 156°C to 160°C
Quantity 1 g
Beilstein 606937
Solubility Information Soluble in dimethyl sulfoxide and methanol. Slightly soluble in water.
Formula Weight 140.14
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Ethyl imidazole-4-carboxylate