Learn More
Ethyl 5-benzyloxyindole-2-carboxylate, 97%, Thermo Scientific Chemicals
Catalog Number p-7044081
Click to view available options
Quantity:
1 g
5 g
37033-95-7
C18H17NO3
295.338
MFCD00022702
DCIFXYFKVKDOLL-UHFFFAOYSA-N
ethyl 5-benzyloxy-1h-indole-2-carboxylate, ethyl 5-benzyloxyindole-2-carboxylate, 5-benzyloxyindole-2-carboxylic acid ethyl ester, 5-benzyloxy-2-carbethoxyindole, ethyl 5-benzyloxy indole-2-carboxylate, 5-benzyloxyindole-2-carboxylic acid ethylester, ethyl 5-phenylmethoxy-1h-indole-2-carboxylate, 5-phenylmethoxy-1h-indole-2-carboxylic acid ethyl ester, 5-benzyloxyindole-2-carboxylic acid*ethyl ester c, 1h-indole-2-carboxylic acid, 5-phenylmethoxy-, ethyl ester
95721
ethyl 5-phenylmethoxy-1H-indole-2-carboxylate
CCOC(=O)C1=CC2=C(N1)C=CC(=C2)OCC3=CC=CC=C3
37033-95-7
C18H17NO3
295.338
MFCD00022702
DCIFXYFKVKDOLL-UHFFFAOYSA-N
ethyl 5-benzyloxy-1h-indole-2-carboxylate, ethyl 5-benzyloxyindole-2-carboxylate, 5-benzyloxyindole-2-carboxylic acid ethyl ester, 5-benzyloxy-2-carbethoxyindole, ethyl 5-benzyloxy indole-2-carboxylate, 5-benzyloxyindole-2-carboxylic acid ethylester, ethyl 5-phenylmethoxy-1h-indole-2-carboxylate, 5-phenylmethoxy-1h-indole-2-carboxylic acid ethyl ester, 5-benzyloxyindole-2-carboxylic acid*ethyl ester c, 1h-indole-2-carboxylic acid, 5-phenylmethoxy-, ethyl ester
95721
ethyl 5-phenylmethoxy-1H-indole-2-carboxylate
CCOC(=O)C1=CC2=C(N1)C=CC(=C2)OCC3=CC=CC=C3
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 160°C to 165°C
Quantity 1 g
Beilstein 281129
Formula Weight 295.34
Percent Purity 97%
Chemical Name or Material Ethyl 5-benzyloxyindole-2-carboxylate