missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-aminophenylacetate, 98%, Thermo Scientific Chemicals

Catalog Number p-7081520
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
5438-70-0
C10H13NO2
179.219
MFCD00017569
CFNDVXUTYPXOPG-UHFFFAOYSA-N
ethyl 4-aminophenylacetate, ethyl 2-4-aminophenyl acetate, p-aminophenylacetic acid ethyl ester, 4-aminophenylacetic acid ethyl ester, ethyl 4-aminophenyl acetate, ethyl p-aminophenylacetate, unii-5h8cfu7gnp, benzeneacetic acid, 4-amino-, ethyl ester, ethyl-4-aminophenylacetate
225219
ethyl 2-(4-aminophenyl)acetate
CCOC(=O)CC1=CC=C(C=C1)N
This item is not returnable. View return policy
5438-70-0
C10H13NO2
179.219
MFCD00017569
CFNDVXUTYPXOPG-UHFFFAOYSA-N
ethyl 4-aminophenylacetate, ethyl 2-4-aminophenyl acetate, p-aminophenylacetic acid ethyl ester, 4-aminophenylacetic acid ethyl ester, ethyl 4-aminophenyl acetate, ethyl p-aminophenylacetate, unii-5h8cfu7gnp, benzeneacetic acid, 4-amino-, ethyl ester, ethyl-4-aminophenylacetate
225219
ethyl 2-(4-aminophenyl)acetate
CCOC(=O)CC1=CC=C(C=C1)N

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 46°C to 48°C
Quantity 5 g
Formula Weight 179.22
Percent Purity 98%
Chemical Name or Material Ethyl 4-aminophenylacetate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'