missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-isothiocyanatopropionate, 97%, Thermo Scientific Chemicals

Catalog Number p-7045487
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
17126-62-4
C6H9NO2S
159.203
MFCD00041130
UPTRONYNXNYITM-UHFFFAOYSA-N
ethyl 3-isothiocyanatopropionate, 3-isothiocyanato-propionic acid ethyl ester, acmc-1ca2j, ethyl-3-isothiocynato propionate, isothiocyanato-propionic acid ethyl ester, 3-isothiocyanatopropionic acid ethyl ester, propanoic acid,3-isothiocyanato-,ethyl ester, propanoic acid,3-isothiocyanato-, ethyl ester
526514
ethyl 3-isothiocyanatopropanoate
CCOC(=O)CCN=C=S
This item is not returnable. View return policy
17126-62-4
C6H9NO2S
159.203
MFCD00041130
UPTRONYNXNYITM-UHFFFAOYSA-N
ethyl 3-isothiocyanatopropionate, 3-isothiocyanato-propionic acid ethyl ester, acmc-1ca2j, ethyl-3-isothiocynato propionate, isothiocyanato-propionic acid ethyl ester, 3-isothiocyanatopropionic acid ethyl ester, propanoic acid,3-isothiocyanato-,ethyl ester, propanoic acid,3-isothiocyanato-, ethyl ester
526514
ethyl 3-isothiocyanatopropanoate
CCOC(=O)CCN=C=S

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.12
Boiling Point 75°C to 76°C (8 mmHg)
Odor Pungent
Linear Formula SCNCH2CH2CO2CH2CH3
Refractive Index 1.5014
Quantity 1 g
UN Number UN2922
Beilstein 1705633
Sensitivity Moisture sensitive
Formula Weight 159.2
Percent Purity 97%
Chemical Name or Material Ethyl 3-isothiocyanatopropionate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'