missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3,4,5-trihydroxybenzoate, 98%, Thermo Scientific Chemicals

Catalog Number p-7021413
423.62 NOK valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
C9H10O5
198.174
MFCD00016430
VFPFQHQNJCMNBZ-UHFFFAOYSA-N
ethyl gallate, gallic acid ethyl ester, phyllemblin, nipagallin a, progallin a, ethylgallate, nipa no. 48, gallic acid, ethyl ester, benzoic acid, 3,4,5-trihydroxy-, ethyl ester, nipa 48
13250
CHEBI:87247
ethyl 3,4,5-trihydroxybenzoate
CCOC(=O)C1=CC(=C(C(=C1)O)O)O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
C9H10O5
198.174
MFCD00016430
VFPFQHQNJCMNBZ-UHFFFAOYSA-N
ethyl gallate, gallic acid ethyl ester, phyllemblin, nipagallin a, progallin a, ethylgallate, nipa no. 48, gallic acid, ethyl ester, benzoic acid, 3,4,5-trihydroxy-, ethyl ester, nipa 48
13250
CHEBI:87247
ethyl 3,4,5-trihydroxybenzoate
CCOC(=O)C1=CC(=C(C(=C1)O)O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 148°C to 152°C
Quantity 10 g
Formula Weight 198.18
Percent Purity 0.98
Chemical Name or Material Ethyl 3,4,5-trihydroxybenzoate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'