missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-bromobenzoate, 98+%, Thermo Scientific Chemicals

Catalog Number p-7021008
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
6091-64-1
C9H9BrO2
229.073
MFCD00015443
BIHHBTVQFPVSTE-UHFFFAOYSA-N
2-bromobenzoic acid ethyl ester, benzoic acid, 2-bromo-, ethyl ester, ethyl-2-bromobenzoate, ethyl2-bromobenzoate, ethyl bromobenzoate, ethyl o-bromobenzoate, pubchem3937, brombenzoesaureathylester, acmc-209mn8, ksc493o4r
80186
ethyl 2-bromobenzoate
CCOC(=O)C1=CC=CC=C1Br
This item is not returnable. View return policy
6091-64-1
C9H9BrO2
229.073
MFCD00015443
BIHHBTVQFPVSTE-UHFFFAOYSA-N
2-bromobenzoic acid ethyl ester, benzoic acid, 2-bromo-, ethyl ester, ethyl-2-bromobenzoate, ethyl2-bromobenzoate, ethyl bromobenzoate, ethyl o-bromobenzoate, pubchem3937, brombenzoesaureathylester, acmc-209mn8, ksc493o4r
80186
ethyl 2-bromobenzoate
CCOC(=O)C1=CC=CC=C1Br

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.44
Boiling Point 255°C
Flash Point >110°C (230°F)
Refractive Index 1.544
Quantity 10 g
Beilstein 2207360
Formula Weight 229.07
Percent Purity ≥98%
Chemical Name or Material Ethyl 2-bromobenzoate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'