missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 2,4-dimethylbenzoate, 98+%, Thermo Scientific™
Click to view available options
Quantity:
5 g
25 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
It is employed as intermediate for pharmaceutical.
Solubility
Not miscible or difficult to mix with water.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
It is employed as intermediate for pharmaceutical.
Solubility
Not miscible or difficult to mix with water.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
Beilstein | 1938737 |
Solubility Information | Not miscible or difficult to mix with water. |
Formula Weight | 178.23 |
Percent Purity | ≥98% |
Quantity | 5 g |
Refractive Index | 1.513 |
Chemical Name or Material | Ethyl 2,4-dimethylbenzoate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'