Learn More
Ethoxyacetic acid, 98%, Thermo Scientific Chemicals
Catalog Number p-7027443
Click to view available options
Quantity:
25 g
100 g
627-03-2
C4H8O3
104.105
MFCD00004310
YZGQDNOIGFBYKF-UHFFFAOYSA-N
ethoxyacetic acid, acetic acid, ethoxy, ethoxy acetic acid, acetic acid, 2-ethoxy, ethoxy-acetic acid, unii-467vw095bx, o-ethylglycolic acid, ccris 7194, 2-ethoxy-acetic acid, dsstox_cid_11294
12301
2-ethoxyacetic acid
CCOCC(=O)O
627-03-2
C4H8O3
104.105
MFCD00004310
YZGQDNOIGFBYKF-UHFFFAOYSA-N
ethoxyacetic acid, acetic acid, ethoxy, ethoxy acetic acid, acetic acid, 2-ethoxy, ethoxy-acetic acid, unii-467vw095bx, o-ethylglycolic acid, ccris 7194, 2-ethoxy-acetic acid, dsstox_cid_11294
12301
2-ethoxyacetic acid
CCOCC(=O)O
Ethoxyacetic acid is used to evaluate toxicity in embryonic stem cell test in mice. It finds application as a biomarker of exposure to alkyl-substituted ethylene glycols and other aliphatic ethers. It is used as a precursor to prepare ethoxy-acetic acid ethoxymethyl ester.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethoxyacetic acid is used to evaluate toxicity in embryonic stem cell test in mice. It finds application as a biomarker of exposure to alkyl-substituted ethylene glycols and other aliphatic ethers. It is used as a precursor to prepare ethoxy-acetic acid ethoxymethyl ester.
Solubility
Miscible with water.
Notes
Incompatible with bases, oxidizing agents and reducing agents.
TRUSTED_SUSTAINABILITY
Density 1.102
Boiling Point 207°C
Flash Point 97°C (206°F)
Odor Pungent
Refractive Index 1.419
Quantity 25 g
UN Number UN3265
Beilstein 1743037
Solubility Information Miscible with water.
Formula Weight 104.11
Percent Purity 98%
Chemical Name or Material Ethoxyacetic acid