missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ ERGIC1 (Human) Recombinant Protein
Human ERGIC1 full-length ORF ( NP_001026881.1, 1 a.a. - 290 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00057222-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MPFDFRRFDIYRKVPKDLTQPTYTGAIISICCCLFILFLFLSELTGFITTEVVNELYVDDPDKDSGGKIDVSLNISLPNLHCELVGLDIQDEMGRHEVGHIDNSMKIPLNNGAGCRFEGQFSINKVPGNFHVSTHSATAQPQNPDMTHVIHKLSFGDTLQVQNIHGAFNALGGADRLTSNPLASHDYILKIVPTVYEDKSGKQRYSYQYTVANKEYVAYSHTGRIIPAIWFRYDLSPITVKYTERRQPLYRFITTICAIIGGTFTVAGILDSCIFTASEAWKKIQLGKMH
Specifications
NP_001026881.1 | |
endoplasmic reticulum-golgi intermediate compartment (ERGIC) 1 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ERGIC1 | |
GST |
57222 | |
Wheat germ expression system | |
25 μg | |
ERGIC-32, ERGIC32, FLJ39864, KIAA1181, MGC14345 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |