Learn More
Epidermal growth factor from Mouse Submaxillary Gland, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
Description
- EGF stimulates the proliferation and differentiation of a wide variety of cell types deriving both from ectoderm and mesoderm.
- In tissue culture, EGF can act to reduce or eliminate the requirement for serum and is often used in conjunction with other media additives and hormones, such as insulin, transferrin, and prostaglandin E1. Cellular metabolic effects of EGF include stimulation of ion fluxes, glucose transport, glycolysis, and synthesis of DNA, RNA and proteins. EGF stimulates the proliferation and differentiation of a wide variety of cell types deriving both from ectoderm and mesoderm.
- In tissue culture, EGF can act to reduce or eliminate the requirement for serum and is often used in conjunction with other media additives and hormones, such as insulin, transferrin, and prostaglandin E1.
- Cellular metabolic effects of EGF include stimulation of ion fluxes, glucose transport, glycolysis, and synthesis of DNA, RNA and proteins.
- Epidermal growth factor or EGF is a growth factor isolated from mouse submaxillary glands that stimulates cell growth, proliferation, and differentiation by binding to its receptor EGFR. EGF is mitogenic for a variety of epidermal and epithelial cells, including fibroblasts, glial cells, mammary epithelial cells, vascular and corneal endothelial cells, bovine granulosa, rabbit chondrocytes, HeLa and SV40-3T3 cells.
Specifications
Specifications
| CAS | 62229-50-9 |
| Color | White |
| Molecular Formula | C257H381N73O83S7 |
| MDL Number | MFCD01634807 |
| Quantity | 100 μg |
| Synonym | EGF, Epidermal cellgrowth factor |
| Solubility Information | EGF, Epidermal cellgrowth factor |
| InChI Key | VBEQCZHXXJYVRD-DGWNDDGFNA-N |
| Molecular Weight (g/mol) | 6045.72 |
| PubChem CID | 56841751 |
| Show More |