Learn More
EpCAM/TROP1 Antibody (158206) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB9601JF635
Product Code. 30171313
  • 5785.00 NOK / 0.10mL
Description
EpCAM/TROP1 Monoclonal antibody specifically detects EpCAM/TROP1 in Human samples. It is validated for Flow Cytometry,ELISA Standard (Matched Pair),ELISA Capture (Matched Antibody Pair),ELISA Detection (Matched Antibody Pair),CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
EpCAM/TROP1
Monoclonal
Janelia Fluor 635
17-1A, 323/A3, ACSTD1, antigen identified by monoclonal AUA1, CD326 antigen, Cell surface glycoprotein Trop-1, chromosome 4, surface marker (35kD glycoprotein), DIAR5, EGP, EGP-2, EGP314, EGP40, EpCAM, epithelial cell adhesion molecule, Epithelial cell surface antigen, Epithelial glycoprotein, Epithelial glycoprotein 314, ESA, GA733-2EGP34, hEGP314, HNPCC8, KS 1/4 antigen, KS1/4, KSAHEA125, M1S2, M4S1Ly74, Major gastrointestinal tumor-associated protein GA733-2, MIC18MH99, MOC31, TACST-1, TACSTD1, TROP1CD326, Tumor-associated calcium signal transducer 1CO-17A
Mouse myeloma cell line NS0-derived recombinant human EpCAM/TROP-1, Extracellular domain
RUO
Primary
Human
Liquid
Flow Cytometry, ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), CyTOF
158206
50mM Sodium Borate
Mouse
0.1 mL
Cancer, Cancer Stem Cells, Tumor Biomarkers
4072
Store at 4°C in the dark.
IgG2b