missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPB41L4A Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | EPB41L4A |
---|---|
Concentration | 0.3 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16885015
|
Proteintech
26596-1-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16875015
|
Proteintech
26596-1-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Members of the band 4.1 protein superfamily, including EPB41L4A, are thought to regulate the interaction between the cytoskeleton and plasma membrane (Ishiguro et al., 2000 ).Specifications
EPB41L4A | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
P52963, Q9HCS5 | |
13824, 64097 | |
EPB41L4A Fusion Protein Ag23923 | |
Primary | |
-20°C | |
EPB41L4A |
0.3 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Band 4.1 like protein 4A, EPB41L4, EPB41L4A, NBL4, Protein NBL4 | |
EPB41L4A | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |