missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ENPP-3/CD203c Antibody (NP4D6) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB5756JF635
This item is not returnable.
View return policy
Description
ENPP-3/CD203c Monoclonal antibody specifically detects ENPP-3/CD203c in Human samples. It is validated for Flow Cytometry,CyTOF
Specifications
| ENPP-3/CD203c | |
| Monoclonal | |
| Janelia Fluor 635 | |
| CD203c, CD203c antigen, dJ1005H11.3 (phosphodiesterase I/nucleotide pyrophosphatase 3), dJ914N13.3 (phosphodiesterase I/nucleotide pyrophosphatase 3), ectonucleotide pyrophosphatase/phosphodiesterase 3, ectonucleotide pyrophosphatase/phosphodiesterase family member 3, E-NPP 3, gp130RB13-6, NPP3, PD-IBETA, PDNP3B10, Phosphodiesterase I beta, Phosphodiesterase I/nucleotide pyrophosphatase 3, phosphodiesterase-I beta | |
| HEK293 human embryonic kidney cell line transfected with human ENPP-3/CD203c, Accession # O14638 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, CyTOF | |
| NP4D6 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Protein Phosphatase | |
| 5169 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction