Learn More
Macherey-Nagel™ Endotoxin-Free Buffers for NucleoBond™ Xtra EF Kits
Click to view available options
:
Buffer: RES-EF, Quantity: 3000 mL
Description
Replacement/extra buffers for use with the kits
Specifications
Specifications
| Chemical Name or Material | RES-EF |
| CAS | 64-17-5d |
| Color | Colorless |
| For Use With (Application) | Endotoxin-free plasmid DNA purification |
| For Use With (Equipment) | NucleoBond™ Xtra EF Kit |
| pH | 7.5 to 8.5 |
| Physical Form | Liquid |
| Quantity | 3000 mL |
| Odor | Odorless |