Learn More
Merck™ Cartridges and Kits for Milli-Q™ Bowl-Style Water Purification Systems
Pretreatment and purification replacement cartridges
Brand:  Merck™ CF0F01205
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10020202
  • 11365.00 NOK / Each
Call for latest availability
+47 22 95 59 59
Add to basket
Description
Description
For Milli-Q™ Bowl systems
Specifications
Specifications
Milli-Q™
Milli-Q™ organicfree ultrapure cartridge kit
Ion exchange, activated Carbon, depth filtration
One Super-C™ organics removal cartridge, two ion exchange deionization cartridges, one Organex-Q trace organic removal cartridge, one Millipak™ filter unit
Milli-Q™ bowl-style water purification system