Learn More
Merck Millipore BioPak™ Disposable Ultrafilter
Nuclease-free, pyrogen-free water
Brand: Merck Millipore CDUFBI001
Additional Details : Weight : 0.20700kg
Description
Nuclease-free, pyrogen-free water
- Guaranteed nuclease-free and pyrogen-free performance for three months, regardless of volumes used
- Disposable filter eliminates the need for regular, lengthy sanitization procesures and hazardous chemicals
- Fits all lab-scale EMD Millipore Ultrapure water systems: Simplicity portable water systems, Synergy™ ultrapure water systems, Direct-Q™ water systems, and all generations of Milli-Q™ water systems
- Preserves water quality as well as performance—no loss in system flowrate
- Cost-effective upgrade for your existing EMD Millipore ultrapure water system
Applications:
Biological techniques requiring nuclease-free and/or pyrogen-free water
Replacement of molecular biology-grade and cell culture-grade bottled water with similar performance
Advanced life science research
Specifications
Ultrafiltration | |
EMD Millipore™ Ultrapure™, Synergy™ Ultrapure™, Direct-Q™ and all generations of Milli-Q™ water systems | |
Ultrafiltration Cartridge |
Production of pyrogen-free and RNase-free water | |
1 Pack |