Learn More
Eighteen element solution for ICP/MS 10mg/L HNO3 matrix, Fisher chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Brand: Fisher Chemical MS 008 050
Additional Details : Weight : 0.05000kg
Description
Eighteen elements solution - Ag, Al, As, Ba, Be, Cd, Cr, Co, Cu, Mn, Ni, Pb Se, Th, Tl, U, V, Zn for ICP/MS 10mg/L HNO3 matrixSpecifications
Eighteen Elements | |
Bottle | |
ICP/MS Calibration Multi-element Standard Solution | |
2% HNO3 |
10mg/L | |
50 mL | |
Ag,Al,As,Ba,Be,Cd,Cr,Co,Cu,Mn,Ni,Pb,Se,Th,Tl,U,V,Zn | |
Liquid |
Safety and Handling
Product Identifier
Hazard Category
Hazard Statement
Precautionary Statement
- Multi-element Standard Solution for ICP. 18 Components in HNO3 2% (MS 008)
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
- P501-Dispose of contents/container to .