missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EIF4EBP2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | EIF4EBP2 |
---|---|
Concentration | 0.25 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16888083
|
Proteintech
11182-1-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16878083
|
Proteintech
11182-1-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a member of the eukaryotic translation initiation factor 4E binding protein family. The gene products of this family bind eIF4E and inhibit translation initiation. However, insulin and other growth factors can release this inhibition via a phosphorylation-dependent disruption of their binding to eIF4E. Regulation of protein production through these gene products have been implicated in cell proliferation, cell differentiation and viral infection.Specifications
EIF4EBP2 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
P70445, Q13542 | |
13688, 1979 | |
EIF4EBP2 Fusion Protein Ag1670 | |
Primary | |
-20°C | |
EIF4EBP2 |
0.25 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
4E BP2, 4EBP2, eIF4E binding protein 2, EIF4EBP2, PHASII | |
EIF4EBP2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |