missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EIF2AK2 Mouse anti-Human, Clone: 2A8A4, Proteintech
Mouse Monoclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | EIF2AK2 |
---|---|
Clone | 2A8A4 |
Concentration | 1 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16888715
|
Proteintech
66646-1-IG-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16878715
|
Proteintech
66646-1-IG-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This antibody is specific to EIF2AK2.
PKR is a 68 kDa protein and an IFN-induced serine/threonine protein kinase consisting of two N-terminal RNA-binding regulatory domains and a functional C-terminal serine/threonine protein kinase domain. Activated by dsRNA produced during viral infections, PKR plays a key role in IFN induced-innate antiviral response, virus-induced apoptosis, cell growth & differentiation. Upon activation, PKR gets autophosphorylated and catalyzes the phosphorylation of elF2 alpha subunit, which leads to an inhibition of protein synthesis. PKR induces apoptosis by up-regulating Fas expression and mediates FADD/Caspase-8 death signaling pathway. Upon viral infection, it functions as a dual protein, sequentially activating both cell survival & cell death pathways using kinase independent and dependent strategies. PRKR regulates multiple pathways which include NF-kappaB activation, p53, p38, & PDGF signaling pathway. PKR has been implicated in tumor suppression & maligcy. To evade the antiviral effects of PKR, viruses have evolved multiple mechanisms, such as the inhibition of PKR by the non-structural protein (NS1) of the influenza virus. More recently, PKR has been implicated in several neurodegenerative diseases including Alzheimer, Huntington, and amyotrophic lateral sclerosis.Specifications
EIF2AK2 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
eIF 2A protein kinase 2, EIF2AK1, EIF2AK2, P1/eIF 2A protein kinase, p68 kinase, PKR, PRKR, Protein kinase RNA activated | |
EIF2AK2 | |
IgG1 | |
Protein G | |
Antibody |
2A8A4 | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Human | |
P19525 | |
5610 | |
EIF2AK2 Fusion Protein Ag13010 | |
Primary | |
-20°C | |
EIF2AK2 |