missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EDNRB/Endothelin R Type B Antibody (671917) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB4496JF525
This item is not returnable.
View return policy
Description
EDNRB/Endothelin R Type B Monoclonal antibody specifically detects EDNRB/Endothelin R Type B in Human samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,CyTOF
Specifications
| EDNRB/Endothelin R Type B | |
| Monoclonal | |
| Janelia Fluor 525 | |
| ABCDS, endothelin B receptor, Endothelin receptor non-selective type, endothelin receptor type B, ETB, ET-B, ETBR, ET-BR, ETRB, HSCR, HSCR2, WS4A | |
| NS0 mouse myeloma cell line transfected with human EDNRB/Endothelin R Type B, Accession # P24530 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, Immunohistochemistry, Western Blot, CyTOF | |
| 671917 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| GPCR, Neuroscience, Signal Transduction | |
| 1910 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction