Learn More
Brady™ Economy Polypropylene Tapes
For use indoors on smooth, flat surfaces. Brady economy polypropylene tapes are used with BMP 71 printer.
445.00 NOK - 565.00 NOK
Specifications
Description | Suggested ribbon series: R4400; R6000; R6900 |
---|---|
Adhesive Type | Acrylic, Permanent |
Color | White |
Finish | Gloss |
Material | Polypropylene (B-529) |
Product Code | Brand | Width (Metric) | Width (English) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Width (Metric) | Width (English) | Price | Quantity & Availability | ||||
11573924
|
Brady™
114710 |
2.54 cm | 1 in. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
11553974
|
Brady™
114760 |
5.08 cm | 2 in. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Specifications
Suggested ribbon series: R4400; R6000; R6900 | |
White | |
Polypropylene (B-529) | |
BMP™71 Printer | |
1 Ea. | |
0.0035 in. | |
50 ft. |
Acrylic, Permanent | |
Gloss | |
Economy polypropylene tape | |
1 | |
-40°C to +100°C | |
0.089 mm | |
15.2 m |