Learn More
Tenak™ ECO AluRack for Upright Freezers, 100 mm Box Height, with Boxes
ECO AluRack fits into any upright freezer and can be used with all standard cryoboxes.
1345.00 NOK - 2255.00 NOK
Specifications
Includes | White cardboard boxes, 9 x 9 cell dividers |
---|---|
Description | Seawater resistant |
Material | Aluminum |
For Use With (Equipment) | Upright Freezers |
Height (Metric) | 100 mm (Box) |
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
12412560
|
Tenak™
TE11310B |
Boxes (10) |
1780.00 NOK
Each |
Estimated Shipment: 10-07-2024
Log in to see stock. |
|||||
12422550
|
Tenak™
TE11294B |
Boxes (6) |
1345.00 NOK
Each |
Estimated Shipment: 10-07-2024
Log in to see stock. |
|||||
12432560
|
Tenak™
TE11312B |
Boxes (15) |
2255.00 NOK
Each |
Estimated Shipment: 09-07-2024
Log in to see stock. |
|||||
12442550
|
Tenak™
TE11296B |
Boxes (9) |
1625.00 NOK
Each |
Estimated Shipment: 09-07-2024
Log in to see stock. |
|||||
12472550
|
Tenak™
TE11302B |
Boxes (8) |
1550.00 NOK
Each |
Estimated Shipment: 10-07-2024
Log in to see stock. |
|||||
12492550
|
Tenak™
TE11304B |
Boxes (12) |
1925.00 NOK
Each |
Estimated Shipment: 09-07-2024
Log in to see stock. |
|||||
Description
- Made of seawater resistant aluminum
- Fits into any freezer and can be used with all standard cryoboxes
- Racks can be used with all freezer brands
- Includes standard white cardboard boxes with 9 x 9 cell dividers
- Holds is expressed by Height x Dept
Specifications
White cardboard boxes, 9 x 9 cell dividers | |
Aluminum | |
100 mm (Box) |
Seawater resistant | |
Upright Freezers |