Learn More
Tenak™ ECO AluRack for Chest Freezers, 100 mm Box Height
ECO AluRack fits into any chest freezer and can be used with all standard cryoboxes.
710.00 NOK - 785.00 NOK
Specifications
Description | Seawater resistant |
---|---|
Material | Aluminum |
For Use With (Equipment) | Chest Frseezers |
Height (Metric) | 100 mm (Box) |
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
12408150
|
Tenak™
TE11560 |
Boxes (3) |
710.00 NOK
Each |
Estimated Shipment: 12-07-2024
Log in to see stock. |
|||||
12428150
|
Tenak™
TE11562 |
Boxes (4) |
735.00 NOK
Each |
Estimated Shipment: 12-07-2024
Log in to see stock. |
|||||
12448150
|
Tenak™
TE11564 |
Boxes (5) |
785.00 NOK
Each |
Estimated Shipment: 12-07-2024
Log in to see stock. |
|||||
Description
- Made of seawater resistant aluminum
- Fits into any freezer and can be used with all standard cryoboxes
- Racks can be used with all freezer brands
- Without standard white cardboard boxes
Specifications
Seawater resistant | |
Aluminum | |
Chest Frseezers | |
100 mm (Box) |