Learn More
Tenak™ Eco AluRack for Chest Freezer, for Microtiterplates, 45 mm Shelf Height
Eco AluRack for chest freezers is designed to hold microtiterplates.
700.00 NOK - 1275.00 NOK
Specifications
Material | Aluminum |
---|---|
For Use With (Equipment) | Chest Frseezers |
Height (Metric) | 45 mm |
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
12408190
|
Tenak™
TE15172 |
Microtiterplates (12) |
1060.00 NOK
Each |
Estimated Shipment: 28-06-2024
Log in to see stock. |
|||||
12438190
|
Tenak™
TE15178 |
Microtiterplates (15) |
1225.00 NOK
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
12448180
|
Tenak™
TE15160 |
Microtiterplates (6) |
700.00 NOK
Each |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
12448190
|
Tenak™
TE15180 |
Microtiterplates (16) |
1275.00 NOK
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
12458180
|
Tenak™
TE15162 |
Microtiterplates (7) |
760.00 NOK
Each |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
12468180
|
Tenak™
TE15164 |
Microtiterplates (8) |
825.00 NOK
Each |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
12478180
|
Tenak™
TE15166 |
Microtiterplates (9) |
880.00 NOK
Each |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
12498180
|
Tenak™
TE15170 |
Microtiterplates (11) |
995.00 NOK
Each |
Estimated Shipment: 28-06-2024
Log in to see stock. |
|||||
Description
- For use in chest freezers
- Racks can either be used as one plate per shelf or the plates can be stacked
- Constructed of aluminum
Specifications
Aluminum | |
Chest Frseezers | |
45 mm |