missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ECM2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21376-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
ECM2 encodes extracellular matrix protein 2, so named because it shares extensive similarity with known extracelluar matrix proteins. Three transcript variants encoding different isoforms have been found for this gene.Specifications
ECM2 | |
Polyclonal | |
Unconjugated | |
ECM2 | |
ECM2, Extracellular matrix protein 2, Matrix glycoprotein SC1, Matrix glycoprotein SC1/ECM2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100361383, 1842, 407800 | |
-20°C | |
Liquid |
Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
0.18 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O94769, Q5FW85 | |
ECM2 | |
ECM2 Fusion Protein Ag15913 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |