Learn More
Trajan Scientific and Medical™ EasyLOK Fitting
Composed of knurled stainless steel nut and a double-ended PEEK ferrule for a leak-free connection.
565.00 NOK - 720.00 NOK
Specifications
For Use With (Equipment) | SGE GLT Columns |
---|---|
Max. Pressure | 5000 psi |
Thread Size | 10-32 |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
15239203
|
Trajan Scientific and Medical™
206102 |
EasyLOK Fitting 2 fittings, 2 ferrules |
565.00 NOK
Pack of 2 |
Estimated Shipment: 19-06-2024
Log in to see stock. |
|||||
15249203
|
Trajan Scientific and Medical™
206160 |
PEEK Double Ended Ferrules |
720.00 NOK
Pack of 10 |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
Description
- Leak free connections
- Replacement parts such as nuts and ferrules
Specifications
SGE GLT Columns | |
5000 psi | |
10-32 |